Wang resin on polystyrene (PS), beads 100-200 mesh Loading: 0.9-1.0 mmol/g
Supplier: Supra Sciences
Synonyms:
4-Benzyloxybenzyl alcohol resin, 4-Alkoxybenzyl alcohol resin, 4-HOCH₂-Ph-O-CH₂-Ph-polymer
Common reagent utilized for peptide synthesis on solid support.
Wang Resin is a gel-type polystyrene resin and is a support for solid phase synthesis of peptide amides by coupling amino acids through esterification. This versatile support is capable of forming amines through carbamate formation/cleavage as well as supported small molecule synthesis when carboxylic acids are required. Racemization has been reported utilizing Wang resins, so during activation caution is suggested to minimize the risk. Fmoc protection strategies are commonly employed with Wang and it is customarily cleaved utilizing high percentages of TFA. G
MDL Number:
MFCD00131778 CAS Number: 201058-08-4 |
|
Specification Test Results
Colour | Off-white |
Form | Granule |
Loading | 0.9-1.0 mmol/g |
Particle size | 100-200 mesh |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...