Trithiocyanuric acid on resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 3.0-3.4 mmol/g

Supplier: Supra Sciences

SPMP26-25 SPMP26-100 SPMP26-1KG
103061-954EA 184.62 USD
103061-954 103061-956 103061-958
Trithiocyanuric acid on resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 3.0-3.4 mmol/g
Trithiocyanuric acid

Trimercaptotriazine resin (TMT resin), highly effective scavenger of organometallic species.


Trimercaptotriazine Resin, MP, is a macroporous polystyrene resin functionalized with a trimercaptotriazine end group while its Silica counterpart is bound to a silica backbone. This reagent has shown to be an exceptional scavenger of organometallic reagents. MP-TMT is also extremely efficient in its ability to scavenge Copper. The limitations of solution phase trimercaptotriazine are based primarily around solubility in various solvents. The macroporous backbone of MP-TMT allows for its use in all solvents, and the Silica counterpart is effective in all solvents except any protic media. Simple filtration removes it (and the complexed metal) from your reaction.

Formula: C₃H₃N₃S₃
MDL Number: MFCD00006052
CAS Number: 638-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49e5f25efa6e23545fa0a13f72d55741615a89a6&vid=9456b481bb2af8f5b8afd5fd68e026a18a80fb1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour Off-white
Form Granule
Loading 3.0-3.4 mmol/g
Particle size 330-1225 μm (15-50 mesh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR