Thiourea resin (SI), beads Avg. 40-62 µm Loading: 0.7-0.9 mmol/g

Supplier: Supra Sciences

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

SPSI21-25 SPSI21-100 SPSI21-1KG
103061-946EA 87.69 USD
103061-946 103061-948 103061-950
Thiourea resin (SI), beads Avg. 40-62 µm Loading: 0.7-0.9 mmol/g
Thiourea

Versatile scavenger of organometallic species.


Thiourea Resin, MP, is a macroporous polystyrene resin functionalized with a thiourea end group. Silica Thiourea is a silica based alkyl thioruea. Both of these reagents are exceptional scavengers of organometallic reagents with the ability to sequester Pd(0) being particularly noteworthy. MP/silica thiourea is also extremely efficient in its ability to scavenge other transition metals such as Pt, Ru, V, Ag and Hg. It has also found use as a reagent for hydrogenolysis of endoperoxides as well as functionality as a catalyst once complexed to a transition metal.

Formula: NH₂CSNH₂
MW: 76.12 g/mol
MDL Number: MFCD00008067
CAS Number: 62-56-6
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b1d4f6c2035bfa97eae0290c820b6f2dfa63ccb&vid=a83862392f98e4a6eaeeb02bd14408ebb1007391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour Off-white
Form Granule
Loading 0.7-0.9 mmolg
Particle size 40-62 μm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR