Thiourea resin (MP), beads, macroporous 100-200 mesh Loading: 1.7-1.8 mmol/g

Supplier: Supra Sciences

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

SPMP17-25 SPMP17-100 SPMP17-1KG
103061-938EA 138.46 USD
103061-938 103061-940 103061-942
Thiourea resin (MP), beads, macroporous 100-200 mesh Loading: 1.7-1.8 mmol/g
Thiourea

Versatile scavenger of organometallic species.


Thiourea Resin, MP, is a macroporous polystyrene resin functionalized with a thiourea end group. Silica Thiourea is a silica based alkyl thioruea. Both of these reagents are exceptional scavengers of organometallic reagents with the ability to sequester Pd(0) being particularly noteworthy. MP/silica thiourea is also extremely efficient in its ability to scavenge other transition metals such as Pt, Ru, V, Ag and Hg. It has also found use as a reagent for hydrogenolysis of endoperoxides as well as functionality as a catalyst once complexed to a transition metal.

Formula: NH₂CSNH₂
MW: 76.12 g/mol
MDL Number: MFCD00008067
CAS Number: 62-56-6
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b1d4f6c2035bfa97eae0290c820b6f2dfa63ccb&vid=a83862392f98e4a6eaeeb02bd14408ebb1007391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour Off-white
Form Granule
Loading 1.7-1.8 mmol/g
Particle size 100-200 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR