Thiourea resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 3.4-3.6 mmol/g
Supplier: Supra Sciences
Synonyms:
Sulfourea, Thiocarbamide, Sulphourea
Versatile scavenger of organometallic species.
Thiourea Resin, MP, is a macroporous polystyrene resin functionalized with a thiourea end group. Silica Thiourea is a silica based alkyl thioruea. Both of these reagents are exceptional scavengers of organometallic reagents with the ability to sequester Pd(0) being particularly noteworthy. MP/silica thiourea is also extremely efficient in its ability to scavenge other transition metals such as Pt, Ru, V, Ag and Hg. It has also found use as a reagent for hydrogenolysis of endoperoxides as well as functionality as a catalyst once complexed to a transition metal.
Formula:
NH₂CSNH₂ MW: 76.12 g/mol |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 Merck Index: 13,09443 |
|
Specification Test Results
Colour | Off-white |
Form | Granule |
Loading | 3.4-3.6 mmol/g |
Particle size | 330-1225 μm (15-50 mesh) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...