Adhesive Vinyl GHS OSHA Labels 'Isooctane', HCL Labels
Supplier: HCL Label
GH10532VN0023 GH10532VN0035 GH10532VN0047 GH20532VN0023 GH20532VN0035 GH20532VN0047 GH30532VN0023 GH30532VN0035 GH30532VN0047 GH40532VN0023 GH40532VN0035 GH40532VN0047
103022-002PK 74.03 USD
103022-002 103022-004 103022-006 103022-008 103022-010 103022-012 103030-244 103030-246 103030-248 103030-250 103030-252 103030-254 103037-868 103037-870 103037-872 103037-874 103037-876 103037-878 103045-768 103045-770 103045-772 103045-774 103045-776 103045-778
Adhesive Vinyl GHS OSHA Labels 'Isooctane', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant