Adhesive Vinyl GHS OSHA Labels '20% Fluorine In Nitrogen', HCL Labels
Supplier: HCL Label
GH10401VN0023 GH10401VN0035 GH10401VN0047 GH20401VN0023 GH20401VN0035 GH20401VN0047 GH30401VN0023 GH30401VN0035 GH30401VN0047 GH40401VN0023 GH40401VN0035 GH40401VN0047
103020-566PK 74.03 USD
103020-566 103020-568 103020-650 103020-652 103020-654 103020-656 103028-582 103028-584 103028-586 103028-588 103028-590 103028-592 103035-904 103035-906 103035-908 103035-910 103035-912 103035-914 103044-596 103044-598 103044-600 103044-602 103044-604 103044-606
Adhesive Vinyl GHS OSHA Labels '20% Fluorine In Nitrogen', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.