Adhesive Vinyl GHS OSHA Labels 'Pad Etch', HCL Labels
Supplier: HCL Label
GH10057VN0023 GH10057VN0035 GH10057VN0047 GH20057VN0023 GH20057VN0035 GH20057VN0047 GH30057VN0023 GH30057VN0035 GH30057VN0047 GH40057VN0023 GH40057VN0035 GH40057VN0047
103019-634PK 74.03 USD
103019-634 103019-636 103019-638 103019-640 103019-642 103019-644 103027-546 103027-548 103027-550 103027-552 103027-554 103027-556 103035-340 103035-342 103035-344 103035-346 103035-348 103035-350 103043-272 103043-274 103043-276 103043-278 103043-280 103043-282
Adhesive Vinyl GHS OSHA Labels 'Pad Etch', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant