Adhesive Vinyl GHS OSHA Labels 'Disilane, HCL Labels
Supplier: HCL Label
GH10486VN0023 GH10486VN0035 GH10486VN0047 GH1X235VN0047 GH1X235VN0023 GH1X235VN0035 GH20486VN0023 GH20486VN0035 GH20486VN0047 GH2X235VN0023 GH2X235VN0035 GH2X235VN0047 GH30486VN0047 GH30486VN0023 GH30486VN0035 GH3X235VN0023 GH3X235VN0035 GH3X235VN0047 GH40486VN0023 GH40486VN0035 GH40486VN0047 GH4X235VN0023 GH4X235VN0035 GH4X235VN0047
103021-670PK 74.03 USD
103021-670 103021-672 103021-674 103021-676 103021-678 103021-680 103025-502 103025-504 103026-278 103026-280 103026-282 103026-284 103029-696 103029-698 103029-700 103029-702 103029-704 103029-706 103034-064 103034-066 103034-068 103034-070 103034-416 103034-418 103037-336 103037-338 103037-560 103037-562 103037-564 103037-566 103041-156 103041-158 103041-160 103041-162 103041-164 103041-166 103045-420 103045-422 103045-424 103045-426 103045-428 103045-430 103050-538 103050-540 103050-542 103050-544 103050-546 103050-548
Adhesive Vinyl GHS OSHA Labels 'Disilane, HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.