Adhesive Vinyl GHS OSHA Labels 'Ethyl Alcohol', HCL Labels
Supplier: HCL Label
GH10256VN0023 GH10256VN0035 GH10256VN0047 GH20256VN0023 GH20256VN0035 GH20256VN0047 GH30256VN0023 GH30256VN0035 GH30256VN0047 GH40256VN0023 GH40256VN0035 GH40256VN0047
103020-290PK 74.03 USD
103020-290 103020-292 103020-294 103020-296 103020-298 103020-300 103028-122 103028-124 103028-126 103028-128 103028-202 103028-204 103036-052 103036-054 103036-056 103036-058 103036-060 103036-062 103044-096 103044-098 103044-100 103044-102 103044-104 103044-106
Adhesive Vinyl GHS OSHA Labels 'Ethyl Alcohol', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.