Adhesive Vinyl GHS OSHA Labels 'Trimethylboron', HCL Labels
Supplier: HCL Label
GH10653VN0023 GH10653VN0035 GH10653VN0047 GH20653VN0023 GH20653VN0035 GH20653VN0047 GH30653VN0023 GH30653VN0035 GH30653VN0047 GH40653VN0023 GH40653VN0035 GH40653VN0047
103023-762PK 74.03 USD
103023-762 103023-764 103023-766 103023-768 103023-770 103023-772 103031-660 103031-662 103031-664 103031-666 103031-668 103031-670 103039-616 103039-618 103039-620 103039-622 103039-624 103039-626 103047-096 103047-098 103047-100 103047-102 103047-104 103047-106
Adhesive Vinyl GHS OSHA Labels 'Trimethylboron', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.