Adhesive Vinyl GHS OSHA Labels 'Sodium Metasilicate', HCL Labels
Supplier: HCL Label
GH10616VN0023 GH10616VN0035 GH10616VN0047 GH20616VN0023 GH20616VN0035 GH20616VN0047 GH30616VN0023 GH30616VN0035 GH30616VN0047 GH40616VN0023 GH40616VN0035 GH40616VN0047
103023-278PK 74.03 USD
103023-278 103023-280 103023-282 103023-284 103023-286 103023-288 103031-256 103031-258 103031-260 103031-262 103031-304 103031-306 103039-080 103039-082 103039-084 103039-086 103039-088 103039-090 103046-692 103046-694 103046-696 103046-698 103046-700 103046-702
Adhesive Vinyl GHS OSHA Labels 'Sodium Metasilicate', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labelling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant