Adhesive Vinyl GHS OSHA Labels 'Silane', HCL Labels
Supplier: HCL Label
GH10044VN0023 GH10044VN0035 GH10044VN0047 GH10603VN0023 GH10603VN0035 GH10603VN0047 GH20044VN0047 GH20044VN0023 GH20044VN0035 GH20603VN0023 GH20603VN0035 GH20603VN0047 GH30044VN0023 GH30044VN0035 GH30044VN0047 GH30603VN0023 GH30603VN0035 GH30603VN0047 GH40044VN0023 GH40044VN0035 GH40044VN0047 GH40603VN0023 GH40603VN0035 GH40603VN0047
103019-300PK 74.03 USD
103019-300 103019-302 103019-304 103019-354 103019-356 103019-358 103023-058 103023-060 103023-062 103023-064 103023-066 103023-068 103026-714 103026-716 103026-718 103027-020 103027-022 103027-024 103030-908 103030-910 103030-912 103030-914 103030-916 103030-918 103035-166 103035-168 103035-170 103035-172 103035-174 103035-176 103038-988 103038-990 103038-992 103038-994 103038-996 103038-998 103043-362 103043-364 103043-366 103043-368 103043-370 103043-372 103046-592 103046-594 103046-596 103046-598 103046-600 103046-602
Adhesive Vinyl GHS OSHA Labels 'Silane', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant.