Adhesive Vinyl GHS OSHA Labels '2-Butoxyethyl Acetate', HCL Labels
Supplier: HCL Label
GH10442VN0023 GH10442VN0035 GH10442VN0047 GH20442VN0023 GH20442VN0035 GH20442VN0047 GH30442VN0023 GH30442VN0035 GH30442VN0047 GH40442VN0023 GH40442VN0035 GH40442VN0047
103020-886PK 74.03 USD
103020-886 103020-888 103020-890 103020-892 103020-894 103020-896 103028-902 103028-904 103029-086 103029-088 103029-090 103029-092 103037-280 103037-282 103037-284 103037-286 103037-288 103037-290 103044-980 103044-982 103044-984 103044-986 103044-988 103044-990
Adhesive Vinyl GHS OSHA Labels '2-Butoxyethyl Acetate', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.