Adhesive Vinyl GHS OSHA Labels 'Sulfuric Acid 56%', HCL Labels
Supplier: HCL Label
GH1X941VN0023 GH1X941VN0035 GH1X941VN0047 GH2X941VN0023 GH2X941VN0035 GH2X941VN0047 GH3X941VN0023 GH3X941VN0035 GH3X941VN0047 GH4X941VN0023 GH4X941VN0035 GH4X941VN0047
103026-442PK 74.03 USD
103026-442 103026-444 103026-446 103026-448 103026-450 103026-452 103034-004 103034-006 103034-280 103034-282 103034-284 103034-286 103042-272 103042-274 103042-276 103042-342 103042-344 103042-346 103050-750 103050-752 103050-754 103050-756 103050-758 103050-760
Adhesive Vinyl GHS OSHA Labels 'Sulfuric Acid 56%', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.