Adhesive Vinyl GHS OSHA Labels '20% Sodium Iodide', HCL Labels
Supplier: HCL Label
GH11862VN0023 GH11862VN0035 GH11862VN0047 GH21862VN0023 GH21862VN0035 GH21862VN0047 GH31862VN0023 GH31862VN0035 GH31862VN0047 GH41862VN0047 GH41862VN0023 GH41862VN0035
103025-438PK 74.03 USD
103025-438 103025-440 103025-442 103025-444 103025-446 103025-448 103033-384 103033-386 103033-388 103033-390 103033-392 103033-394 103040-716 103040-718 103040-720 103040-722 103040-724 103040-726 103048-284 103048-286 103048-476 103048-478 103048-480 103048-482
Adhesive Vinyl GHS OSHA Labels '20% Sodium Iodide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.