Adhesive Vinyl GHS OSHA Labels 'Limonene', HCL Labels
Supplier: HCL Label
GH11773VN0047 GH11773VN0023 GH11773VN0035 GH21773VN0023 GH21773VN0035 GH21773VN0047 GH31773VN0023 GH31773VN0035 GH31773VN0047 GH41773VN0023 GH41773VN0035 GH41773VN0047
103025-306PK 102.31 USD
103025-306 103025-308 103025-418 103025-420 103025-422 103025-424 103033-220 103033-222 103033-224 103033-226 103033-228 103033-230 103041-736 103041-738 103041-740 103041-742 103041-744 103041-746 103048-384 103048-386 103048-388 103048-390 103048-392 103048-394
Adhesive Vinyl GHS OSHA Labels 'Limonene', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.