Adhesive Vinyl GHS OSHA Labels 'Phosphoric Acid 85%', HCL Labels
Supplier: HCL Label
GH11076VN0023 GH11076VN0035 GH11076VN0047 GH21076VN0023 GH21076VN0035 GH21076VN0047 GH31076VN0023 GH31076VN0035 GH31076VN0047 GH41076VN0023 GH41076VN0035 GH41076VN0047
103024-070PK 74.03 USD
103024-070 103024-072 103024-090 103024-092 103024-094 103024-096 103032-032 103032-034 103032-036 103032-038 103032-040 103032-042 103039-206 103039-548 103039-550 103039-552 103039-554 103039-556 103047-404 103047-406 103047-408 103047-410 103047-412 103047-414
Adhesive Vinyl GHS OSHA Labels 'Phosphoric Acid 85%', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.