Adhesive Vinyl GHS OSHA Labels 'Methyl Alcohol', HCL Labels
Supplier: HCL Label
GH10552VN0023 GH10552VN0035 GH10552VN0047 GH20552VN0023 GH20552VN0035 GH20552VN0047 GH30552VN0023 GH30552VN0035 GH30552VN0047 GH40552VN0023 GH40552VN0035 GH40552VN0047
103022-194PK 74.03 USD
103022-194 103022-196 103022-198 103022-200 103022-266 103022-268 103030-548 103030-550 103030-552 103030-554 103030-556 103030-558 103038-572 103038-574 103038-576 103038-578 103038-580 103038-582 103046-040 103046-042 103046-044 103046-046 103046-048 103046-050
Adhesive Vinyl GHS OSHA Labels 'Methyl Alcohol', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant