Adhesive Vinyl GHS OSHA Labels '2-Ethoxyethyl Acetate', HCL Labels
Supplier: HCL Label
GH10492VN0023 GH10492VN0035 GH10492VN0047 GH20492VN0023 GH20492VN0035 GH20492VN0047 GH30492VN0023 GH30492VN0035 GH30492VN0047 GH40492VN0023 GH40492VN0035 GH40492VN0047
103021-702PK 74.03 USD
103021-702 103021-704 103021-706 103021-708 103021-710 103021-712 103029-768 103029-770 103029-772 103029-774 103029-776 103029-778 103037-568 103037-570 103037-572 103037-574 103037-576 103037-578 103045-444 103045-446 103045-448 103045-450 103045-452 103045-454
Adhesive Vinyl GHS OSHA Labels '2-Ethoxyethyl Acetate', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.