Adhesive Vinyl GHS OSHA Labels 'Metal Etch', HCL Labels
Supplier: HCL Label
GH10055VN0023 GH10055VN0035 GH10055VN0047 GH20055VN0023 GH20055VN0035 GH20055VN0047 GH30055VN0023 GH30055VN0035 GH30055VN0047 GH40055VN0023 GH40055VN0035 GH40055VN0047
103019-622PK 74.03 USD
103019-622 103019-624 103019-626 103019-628 103019-630 103019-632 103027-534 103027-536 103027-538 103027-540 103027-542 103027-544 103035-224 103035-226 103035-228 103035-230 103035-336 103035-338 103043-260 103043-262 103043-264 103043-266 103043-268 103043-270
Adhesive Vinyl GHS OSHA Labels 'Metal Etch', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant