ActiveMax* Recombinant Mouse VEGF164
Supplier: ACRO Biosystems
ActiveMax* Recombinant VEGF164 (HPLC-verified), Host: HEK293 cells, Species Reactivity: Mouse, Purity: >95%(SDS-PAGE), Molecular Characterization: Tag Free, with calculated MW of of 19.3 KDa, Synonyms: VEGF164, VEGFA, VPF, Storage: 4 Degree C, Size: 50ug