Masterflex® L/S 24, Pump Tubing, C-Flex®, Cole Parmer
Supplier: OAKTON INSTRUMENTS MS
EW-06427-24
103012-108EA 1611.08 USD
103012-108
Masterflex® L/S 24, Pump Tubing, C-Flex®, Cole Parmer
Tubing Transfer Tubing
It is applicable for general-purpose lab requiring high performance and long life.
  • Biocompatible
  • Optically inspected and factory tested
  • Opaque, white
  • Low gas permeability
Temperature range: –45 to 135 °C