Calcein blue AM

Supplier: Anaspec

AS-89205
103011-232EA 133 USD
103011-232
Calcein blue AM
Calcein blue AM
Formula: C₂₁H₂₃NO₁₁
Storage Temperature: Freezer
CAS Number: 168482-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR