bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 20 mM in water ≥95% (by HPLC, TLC)

Supplier: Anaspec

AS-83218
103011-142EA 153.46 USD
103011-142
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 20 mM in water ≥95% (by HPLC, TLC)
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

Cell-permeant DNA minor groove binding dye


  • Spectral properties: Abs/Em = 350/461 nm
  • Appearance: Colorless solution
  • Storage Temperature: 4°C protected from light
  • Shipping condition: Blue ice

Formula: C₂₇H₂₈N₆O·3HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012679
CAS Number: 23491-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR