(4R)-4-i-Propyl-1,2,3-oxathiazolidine-2,2-dioxide-3-carboxylic acid t-butyl ester ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(4R)-2,2-Dioxido-4-isopropyl-1,2,3-oxathiazolidine, N-BOC protected
16-3010-1G
102995-174EA
348.18
USD
102995-174
(4R)-4-i-Propyl-1,2,3-oxathiazolidine-2,2-dioxide-3-carboxylic acid t-butyl ester ≥97%
(4R)-4-i-Propyl-1,2,3-oxathiazolidine-2,2-dioxide-3-carboxylic acid t-butyl ester
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...