Mouse Tissue-type Plasminogen Activator (tPA) Total Antigens ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
TPN19-K01
102995-046EA 1562.69 USD
102995-046
Mouse Tissue-type Plasminogen Activator (tPA) Total Antigens ELISA , Eagle Biosciences
Assays ELISAs
Mouse Tissue-type Plasminogen Activator (tPA) Total Antigen ELISA detects total tPA in mouse sample.
  • Dynamic Range: 0.1 - 50 ng/ml
  • Sensitivity: 0.1 ng/ml
  • Incubation: 2 hours
The Mouse Tissue-type Plasminogen Activator (tPA) Total Antigens ELISA is intended for the quantitative determination of total plasminogen activator Antigens in biological fluids. Tissue plasminogen activator (tPA) is a serine protease that converts plasminogen to the active serine protease plasmin in the blood fibrinolytic system1,2. It also plays an important role in the removal of incipient thrombi3. tPA is widely used for the thrombolytic treatment of acute myocardial infarction3.