Anti-LKM1 (Liver-Kidney Microsomes) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4053
102994-762EA 817.31 USD
102994-762
Anti-LKM1 (Liver-Kidney Microsomes) ELISA, Eagle Biosciences
Assays ELISAs
Anti-LKM1 ELISA determines IgG antibodies to cytochrome P450 IID6 in human serum or plasma.
  • Dynamic Range: 3 -300 U/ml
  • Sensitivity: 3 U/ml
  • Incubation: 90 minutes
The Anti-LKM1 (Liver-Kidney Microsomes) ELISA Assay Kit is used for the quantitative deter­mination of IgG antibodies to cytochrome P450 IID6 in human serum or plasma.