Anti-3-Hydroxyanthranilic acid Rabbit Polyclonal Antibody

Supplier: Eagle Biosciences
IS1008
102994-546EA 1333.85 USD
102994-546
Anti-3-Hydroxyanthranilic acid Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: 3-HydroxyAnthranilic acid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR