Vitamin B6 HPLC Column, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IC2100RP
102994-496EA 1235.77 USD
102994-496
Vitamin B6 HPLC Column, Eagle Biosciences, Inc.
Chromatography Columns
Vitamin B6 HPLC assay kit column can be used for about 500 to 1000 samples.
  • For use with Vitamin B6 HPLC assay kit
The vitamins pyridoxin, pyridoxal and pyridoxamin and the appropriate phosphate products are summarized as vitamin B6. All forms can be transformed into the active form pyridoxal-5-phosphate. Vitamin B6 functions as a coenzyme and is essential for more than 50 reactions in the protein, carbohydrate and fatty acid metabolism thereby synthesizing, transforming or degrading amino acids. In protein metabolism vitamin B6 supports the resorption of amino acids and their transport into the cells. Furthermore vitamin B6 contributes to the synthesis of neurotransmitters and amine products (histamine).