GLP-1 Total ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
GP131-K01
102994-424EA 1006.92 USD
102994-424
GLP-1 Total ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
GLP-1 Total ELISA determines sum of glucagon-like peptide-1 (7-36) & (9-36) in plasma.
  • Dynamic Range: 2 -54 pmol/L
  • Sensitivity: 0.6 pmol/L
  • Incubation: overnight
The primary amino acid sequence of GLP-1 peptide is identical among mammalian species, i.e. rat, mouse, pig, human, etc. The Eagle Biosciences GLP-1 Total ELISA Assay Kit (enzyme-linked immunoassay kit) is intended quantitative determination of sum value of glucagon-like peptide-1 (7-36) and (9-36) [GLP-1 (7-36)] and (9-36) [GLP-1 (9-36)] in plasma samples. Eagle Biosciences GLP-1 Total ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.