Anti-CYP450 3A4 Rabbit Polyclonal Antibody

Supplier: Eagle Biosciences
CYP30-A100
102994-146EA 555.77 USD
102994-146
Anti-CYP450 3A4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: CYP450 3A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR