Complement MBL pathway ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
COMPLMP320
102994-110EA 1209.62 USD
102994-110
Complement MBL pathway ELISA, Eagle Biosciences, Inc.
Assays ELISAs
The Complement MBL Pathway ELISA is to determine MBL complement pathway in human serum.
  • Sensitivity: Cut-off
  • Dynamic Range: Cut-off
  • Incubation Time: 2 hours
The complement system plays an essential role in chronic, autoimmune and infectious disease. There are three pathways of complement activation, namely the classical, the alternative and the recently discovered MBL pathway.