Celiac EmA (Anti-Endomysium Antibody) IgG ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4045
102994-092EA 765 USD
102994-092
Celiac EmA (Anti-Endomysium Antibody) IgG ELISA, Eagle Biosciences
Assays ELISAs
Celiac EmA IgG ELISA determines IgG autoantibodies to sample endomysial autoantigens.
  • Dynamic Range: 1 - 300 U/ml
  • Sensitivity: 1 U/ml
  • Incubation: 2 hours
Several autoAntigenss have been discussed for celiac disease and Dermatitis herpetiformis recently. Dieterich et al. have described two non-collagenous proteins with an apparent molecular weight of 90 and 300 kDa being the main autoAntigenss detected by EmA (1). Celiac disease, or gluten-sensitivity, is found already in neonates and is characterized by small intestinal damages leading to a so-called “flat” mucosa. Due to this extensive lesions mal-absorption occurs frequently accompanied with a depletion of key nutrients.