Human CD86 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C8631-K01
102994-076EA 869.62 USD
102994-076
Human CD86 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD86 ELISA is to be used for in-vitro quantitative determination CD86 in biological samples.
  • Dynamic Range: 1.187 U/ml - 38 U/ml
  • Sensitivity: 0.6 U/ml
  • Incubation: 2.75 hours
The Eagle Biosciences Human CD86 ELISA is to be used for the in-vitro quantitative determination CD86 in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD86. The Eagle Biosciences Human CD86 ELISA Assay kit is for Research Use Only.