Human CD141 / THROMBOMODULIN ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C4131-K01
102994-054EA 869.62 USD
102994-054
Human CD141 / THROMBOMODULIN ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
sCD141 ELISA is to detect sCD141 in supernatants, buffered solutions or serum and plasma samples.
  • Dynamic Range: 0.625 - 20 ng/ml
  • Sensitivity: 0.31 ng/ml
  • Incubation: 1.75 hours
The Eagle Biosciences sCD141 ELISA kit is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of sCD141 in supernatants, buffered solutions or serum and plasma samples. This assay will recognise both natural and recombinant human sCD141. The Eagle Biosciences Human sCD141 ELISA Assay kit is for Research Use Only.