Human IL-3 ELISA KIT, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IL331-K01
102993-494EA 693.08 USD
102993-494
Human IL-3 ELISA KIT, Eagle Biosciences, Inc.
Assays ELISAs
Human IL-3 ELISA detects human Interleukin 3 (IL-3) concentrations in various sample types.
  • Dynamic Range: 31.25 - 1000
  • Sensitivity: 15 pg/ml
  • Incubation: 3.5 hours
The Eagle Biosciences Human Interleukin 3 (IL-3) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Interleukin 3 (IL-3) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Interleukin 3 (IL-3) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.