Mouse IgE ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
IGE11-K01
102993-444EA 686.54 USD
102993-444
Mouse IgE ELISA KIT, Eagle Biosciences
Assays ELISAs
Mouse IgE ELISA detects mouse IgE concentrations in cell culture supernates, serum, and plasma.
  • Dynamic Range: 312.5 - 10,000 pg/ml
  • Sensitivity: 156 pg/ml
  • Incubation: 3.0 hours
There are five classes of mammalian immunoglobulins: IgA, IgD, IgE, IgG, and IgM. In mice, the IgG class is further divided into four subclasses: IgG1, IgG2a/ IgG2c (strain specific), IgG2b, and IgG3.The general immunoglobulin structure is described as two heavy and two light chain (H2L2) polypeptides linked together by disulfide bridges of cysteine residues. Mouse IgE, also known as reaginic antibody, exists in serum as a 4-chain monomer of 185–200 kDa. Like IgM, the IgE monomers contain a fourth constant region domain. Serum concentrations of IgE are reported to be >10 ug/ml. IgE is cyolytic for mast cells, where it binds to the FcRI receptor and is able to mediate the response of granular and lipid mediators of inflammation, including the PCA reaction. The IgE class switch is induced by IL-4 as part of the Th2 response.