GSK2606414 ≥95%

Supplier: 0000027161

Synonyms: 1-(5-(4-Amino-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-5-yl)indolin-1-yl)-2-(3-(trifluoromethyl)phenyl)ethanone

SYN-1201-M001 SYN-1201-M005 SYN-1201-M010 SYN-1201-M050
102992-754EA 134.31 USD
102992-754 102992-756 102992-758 102992-760
GSK2606414 ≥95%
GSK2606414

GSK2606414 is an orally available, potent, and selective PERK inhibitor with IC(50) of 0.4nM, displaying at least 100-fold selectivity over the other EIF2AKs assayed.

Formula: C₂₄H₂₀F₃N₅O
Storage Temperature: Freezer
MDL Number: MFCD25976926
CAS Number: 1337531-36-8

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR