GDC-046 ≥95%

Supplier: 0000027161

Synonyms: 2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide

SYN-1198-M001 SYN-1198-M005 SYN-1198-M010 SYN-1198-M050
102992-730EA 134.31 USD
102992-730 102992-732 102992-734 102992-736
GDC-046 ≥95%
GDC-046

GDC046 is a potent selective TYK2 inhibitor (Ki=4.8nm) versus JAK1 (Ki=84nm) and JAK2 (Ki=28nm). It is a lead analog with good kinase selectivity, physicochemical properties, and pharmacokinetic profile.

Formula: C₁₆H₁₃Cl₂N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD28155087
CAS Number: 1258292-64-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR