TAE684 ≥95%
Supplier: 0000027161
Synonyms:
5-Chloro-N4-(2-(isopropylsulphonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine, NVP-TAE 684, NVP-TAE684, 5-Chloro-N4-(2-(isopropylsulfonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine
SYN-1159-M001
SYN-1159-M005
SYN-1159-M010
SYN-1159-M050
102992-448EA
134.31
USD
102992-448
102992-450
102992-452
102992-454
TAE684 ≥95%
TAE684
TAE684 is a potent and selective ALK inhibitor with an IC(50) of 3nM. It is 100-fold more sensitive for ALK over InsR.
Formula:
C₃₀H₄₀ClN₇O₃S Storage Temperature: Freezer |
MDL Number:
MFCD11977634 CAS Number: 761439-42-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...