TAK-285 ≥95%

Supplier: 0000027161

Synonyms: N-(2-(4-((3-Chloro-4-(3-(trifluoromethyl)phenoxy)phenyl)amino)-5H-pyrrolo[3,2-d]pyrimidin-5-yl)ethyl)-3-hydroxy-3-methylbutanamide

SYN-1153-M001 SYN-1153-M005 SYN-1153-M010
102992-406EA 246.46 USD
102992-406 102992-408 102992-410
TAK-285 ≥95%
TAK-285

TAK-285 is a novel investigational dual HER2/EGFR inhibitor with IC(50) values of 17 and 23nM, respectively.

Formula: C₂₆H₂₅ClF₃N₅O₃
Storage Temperature: Freezer
MDL Number: MFCD22124520
CAS Number: 871026-44-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR