NIBR-17 ≥95%

Supplier: 0000027161

Synonyms: N6-(6-Methoxypyridin-3-yl)-2-morpholino-[4,5'-bipyrimidine]-2',6-diamine

SYN-1145-M001 SYN-1145-M005 SYN-1145-M010 SYN-1145-M050
102992-350EA 254.77 USD
102992-350 102992-352 102992-354 102992-356
NIBR-17 ≥95%
NIBR-17

NIBR-17 is a pan class I PI3K inhibitor with IC(50) values of 1, 9.2, 9 and 20nM, respectively for PI3Kalpha - delta.

Formula: C₁₈H₂₀N₈O₂
Storage Temperature: Freezer
MDL Number: MFCD22631503
CAS Number: 944396-88-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR