Ponatinib ≥95%

Supplier: 0000027161

Synonyms: 3-(2-Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-benzamide, 3-(Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide, AP24534

SYN-1116-M001 SYN-1116-M005 SYN-1116-M010 SYN-1116-M050
102992-146EA 90 USD
102992-146 102992-148 102992-150 102992-152
Ponatinib ≥95%
Ponatinib

Ponatinib is a potent multi-target inhibitor of Abl, PDGFRalpha, VEGFR2, FGFR1 and Src with IC(50) values of 0.37, 1.1, 1.5, 2.2 and 5.4nM respectively.

Formula: C₂₉H₂₇F₃N₆O
Storage Temperature: Freezer
MDL Number: MFCD17215203
CAS Number: 943319-70-8

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR