A-674563 hydrochloride ≥95%
Supplier: 0000027161
|
Warning
|
SYN-1110-M001
SYN-1110-M005
SYN-1110-M010
102992-106EA
261.69
USD
102992-106
102992-108
102992-110
A-674563 hydrochloride ≥95%
A-674563 hydrochloride
A-674563 is a potent, orally available Akt1 inhibitor with an IC(50) of 11nM. It also displays activity against PKA and CDK2 with IC(50) values of 16 and 46nM respectively.
Formula:
C₂₂H₂₂N₄O·HCl Storage Temperature: Freezer |
CAS Number:
552325-73-2 |
Specification Test Results
Purity | >95% |
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...