Vandetanib ≥95%

Supplier: 0000027161
Warning

Synonyms: ZD6474, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine

SYN-1090-M001 SYN-1090-M005 SYN-1090-M010 SYN-1090-M050
102991-964EA 90 USD
102991-964 102991-966 102991-968 102991-970
Vandetanib ≥95%
Vandetanib

Vandetanib is a tyrosine kinase inhibitor that targets vascular endothelial growth factor receptor (VEGFR, IC(50) of 40nM) and the epidermal growth factor receptor (EGFR).

Formula: C₂₂H₂₄BrFN₄O₂
Storage Temperature: Freezer
MDL Number: MFCD07772346
CAS Number: 443913-73-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR