BMS-387032 hydrochloride ≥95%

Supplier: 0000027161

Synonyms: SNS-032

SYN-1080-M001 SYN-1080-M005 SYN-1080-M010 SYN-1080-M050
102991-884EA 96.92 USD
102991-884 102991-886 102991-888 102991-890
BMS-387032 hydrochloride ≥95%
BMS-387032 hydrochloride

BMS387032 is a novel cyclin-dependent kinase inhibitor, which exhibits potent and selective inhibitory activity against Cdk2, Cdk7, and Cdk9. It inhibits CDK2 with an IC(50) of 48nM and up to 20-fold selective over CDK1/CDK4. It also inhibits CDK7/9 with IC(50) values of 62nM and 4nM. It has little effect on CDK6.

Formula: C₁₇H₂₄N₄O₂S₂·HCl
Storage Temperature: Freezer
CAS Number: 345627-90-9

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR