SB202190 ≥95%

Supplier: 0000027161
Warning

SYN-1073-M001 SYN-1073-M005 SYN-1073-M010 SYN-1073-M050
102991-824EA 74.77 USD
102991-824 102991-826 102991-828 102991-830
SB202190 ≥95%
SB202190

SB202190 is potent and selective inhibitor of the p38alpha and beta, with IC(50) values of 50 and 100nM respectively. It is a powerful regulator of LPS-induced mRNAs in monocytes.

Formula: C₂₀H₁₄FN₃O
Storage Temperature: Freezer
MDL Number: MFCD00941964
CAS Number: 152121-30-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR