Bisindoylmaleimide X hydrochloride ≥95%

Supplier: 0000027161

Synonyms: Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride

SYN-1021-M001 SYN-1021-M005 SYN-1021-M010 SYN-1021-M050
102991-408EA 186.92 USD
102991-408 102991-410 102991-412 102991-414
Bisindoylmaleimide X hydrochloride ≥95%
Bisindoylmaleimide X hydrochloride

Bisindoylmaleimide X is a potent and selective protein kinase C (PKC) inhibitor (IC(50) of 15nM for rat brain PKC). It also inhibits Fas-mediated apoptosis and T cell-mediated autoimmune diseases.

Formula: C₂₆H₂₄N₄O₂·HCl
Storage Temperature: Freezer
MDL Number: MFCD04972010
CAS Number: 145317-11-9

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR