Zinquin ethyl ester ≥99%
Supplier: 0000027161
|
Warning
|
Synonyms:
2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester, 2-[[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester
CDX-Z0013-M001
CDX-Z0013-M005
CDX-Z0013-M050
102991-254EA
105.23
USD
102991-254
102991-256
102991-258
Zinquin ethyl ester ≥99%
Zinquin ethyl ester
Zinc(II) specific fluorophores probes.
Formula:
C₂₁H₂₂N₂O₅S Storage Temperature: Freezer |
CAS Number:
181530-09-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. Slightly soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...